Learn More
Thermo Scientific Chemicals Besifloxacin hydrochloride
Product Code. 17231206 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. Quantity unitSize
17231206 500 mg 500mg
17221206 100 mg 100mg
2 options
Product Code. 17231206
Brand: Thermo Scientific Chemicals 467505000
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 405165-61-9
Molecular Formula C19H22Cl2FN3O3
Specifications
Quantity 500 mg
Formula Weight 430.31
InChI Key PMQBICKXAAKXAY-UHFFFAOYNA-N
SMILES Cl.NC1CCCCN(C1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1Cl)C(O)=O
IUPAC Name 7-(3-aminoazepan-1-yl)-8-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
Molecular Weight (g/mol) 430.30
Chemical Name or Material Besifloxacin hydrochloride