missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCR Antibody (1E11G12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
1999.20 NOK - 5046.30 NOK
Specifications
| Antigen | BCR |
|---|---|
| Clone | 1E11G12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18278323
|
Novus Biologicals
NBP2-61707 |
0.1 mL |
5335.00 NOK 5046.30 NOK / 0.10mL Save 288.70 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628576
|
Novus Biologicals
NBP2-61707-0.025ml |
0.025 mL |
2115.00 NOK 1999.20 NOK / 0.03mL Save 115.80 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCR Monoclonal specifically detects BCR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| BCR | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| 613 | |
| Purified recombinant fragment of human BCR (AA: 139-280) expressed in E. Coli. | |
| Primary | |
| 142.8 kDa |
| 1E11G12 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| ALLD22S11, BCR1, breakpoint cluster region, breakpoint cluster region protein, CMLEC 2.7.11.1, D22S662, PHLFLJ16453, Renal carcinoma antigen NY-REN-26 | |
| BCR | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title