missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCR Antibody (1E11G12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61707
This item is not returnable.
View return policy
Description
BCR Monoclonal specifically detects BCR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| BCR | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| ALLD22S11, BCR1, breakpoint cluster region, breakpoint cluster region protein, CMLEC 2.7.11.1, D22S662, PHLFLJ16453, Renal carcinoma antigen NY-REN-26 | |
| Mouse | |
| 142.8 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 1E11G12 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| BCR | |
| Purified recombinant fragment of human BCR (AA: 139-280) expressed in E. Coli. | |
| Protein G purified | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 613 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction