missing translation for 'onlineSavingsMsg'
Learn More

BCR Antibody (1E11G12) - BSA Free, Novus Biologicals™

Product Code. 18628576 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.10mL
Product Code. Quantity unitSize
18628576 0.025 mL 0.03mL
18278323 0.1 mL 0.10mL
2 options
This item is not returnable. View return policy

Product Code. 18628576

Brand: Novus Biologicals NBP2617070.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BCR Monoclonal specifically detects BCR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCR
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 1E11G12
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias ALLD22S11, BCR1, breakpoint cluster region, breakpoint cluster region protein, CMLEC 2.7.11.1, D22S662, PHLFLJ16453, Renal carcinoma antigen NY-REN-26
Gene Symbols BCR
Host Species Mouse
Immunogen Purified recombinant fragment of human BCR (AA: 139-280) expressed in E. Coli.
Molecular Weight of Antigen 142.8 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 613
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.