Learn More
Axygen™ 165 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
Shop All Axygen Scientific Products
Click to view available options
Includes:
w/Bottom Label
w/out Bottom Label
Unit Size:
Pack of 4800
Description
- Tip format: 96 tip
- Tip volume range: 20 μL to 250 μL
- Tip material: Clear polypropylene
- RNase-/DNase-/human gDNA-free
- Nonpyrogenic
- Available options:
- With or without aerosol-resistant filters
- Nonsterile or sterile
- Standard or Axygen™ Maxymum Recovery™ surface - Compatible heads: 96LT, 8LT
- Rack format: ANSI/SLAS
- Rack material: Polypropylene
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
- Certifications and Seals of Approval
Specifications
Specifications
Volume (Metric) | 165 μL |
Filtered | Filtered |
Sterility | Sterile |
Color | Clear |
Certifications/Compliance | ANSI |
Clean Claims | DNase, Rnase Free |
For Use With (Application) | Non-pyrogenic, used for stringent specifications under strict process controls |
Format | Rack |
Tip Style | Automation, Round |
Packaging | SLAS Rack |
Show More |
Product Content Correction
Product Title
Axygen™ 165 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format >