Learn More
Invitrogen™ Attune™ NxT Software 21 CFR Part 11
Click to view available options
Quantity:
1 License
1 Server license, 10 users
1 Server license, 5 users
Unit Size:
Each
Description
- The software helps prevent unauthorized access to the system and is available for use on computers with the Windows 10 operating system.
- Authentication of user log-in information
- Ability to set password guidelines and expiry dates
- Lockout access and re-authentication requirements after a period of inactivity
- System lockout upon unauthorized attempts to access software
- Ability of administrator to define user roles and permissions
- Identification of data tampering
- Generation of audit trail log
- Ability of administrator to determine functions required to input reason for change
- Tracking of changes to user roles, account management, and system settings
- Assignment of unique identifiers for experiments and objects within experiments to identify any changes from original record
- Audit trail record presented in human-readable format
- Administrator determination of e-signature requirements
- Ability to review records and sign electronically
- Along with the 21 CFR part 11 software license, our service and support organization offers these services to help you validate you instrument, software, and computer: IQOQ and computer software validation services.
Specifications
Specifications
| For Use With (Equipment) | Attune™ NxT Acoustic Focusing Cytometer |
| Product Line | Attune™ |
| Product Type | Flow Cytometer Software |
| Quantity | 1 Server license, 10 users |
| Shipping Condition | Room Temperature |