Learn More
BINDER™ APT-COM™ 4 Series
Product Code. 16484479 Shop All BINDER Products
Click to view available options
Includes:
4 BASIC-Edition Software
4 GLP-Edition Software
4 PROFESSIONAL-Edition Software
Unit Size:
Each
Product Code. 16484479
Brand: BINDER™ 90530040
Multi Management Software provides management, logging, programming, and documentation options. With features ranging from GLP-compliant logging to convenient program creation and management of up to 100 units, the APT-COM™ 4 can be used in all industries and makes complex work processes much easier.
APT-COM™ 4 BASIC Edition:
  • Administration of up to five connected units
  • Log management (creating, deleting, archiving)
  • Documentation of measured values
  • Central overview of all units in both diagram and table form
  • Graphical presentation of measured values
  • Graphical/numerical program editor
  • Manual export of measured values (CSV/PDF file)
  • Multilingual user interface (German, English, French, Spanish, Italian)
  • Optional program execution via APT-COM™
  • Timer function
  • Import of data from APT-COM™ 3
APT-COM™ 4 PROFESSIONAL Edition:
All the same functions as the BASIC Edition and:
  • Administration of up to 100 connected units at the same time
  • Central overview of all units in a room plan
  • Email-based alarm system
  • Web server for displaying results
  • User administration for multiple users and authorizations
  • Automatic, time-managed export of measured values
  • Reports for further processing/archiving
  • Automatic/manual backup and recovery
APT-COM™ 4 GLP Edition
All the same functions as the PROFESSIONAL Edition and:
  • Can be validated according to 21 CFR Part 11 including validation documents
  • Documentation of measured values in a tamper-proof file in accordance with GLP/GMP guidelines on audit trails
  • Electronic signatures
For Use With: BINDER Chambers Series BD, BF, CB-S, CB, CBF, ED, FD, FED, FP, M, KB, KB ECO, KT, KBF, KBF-S, KBF-S ECO, KBF P, KBF LQC, KBW, KBWF, UFV, VD, VDL
Specifications
Product Type Data Collection Software
Includes 4 PROFESSIONAL-Edition Software
Description APT-COM™ 4 PROFESSIONAL-Edition - convenient unit and user management built on the BASIC edition. Suitable for networking up to 100 units.
For Use With (Application) Logging, Controlling, Monitoring
For Use With (Equipment) BINDER Chambers Series (BD, BF, CB-S, CB, CBF, ED, FD, FED, FP, M, KB, KB ECO, KT, KBF, KBF-S, KBF-S ECO, KBF P, KBF LQC, KBW, KBWF, UFV, VD, VDL)
Operating System Microsoft Windows 10 IoT 64-bit
Product Line APT-COM