Learn More
Fisherbrand™ Trays and Dividers for Shell Vials
Product Code. 10149701 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Type:
Tray for Wide Vials
For Use With (Equipment):
For wide vials (28.5 x 95 mm)
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
10149701 Tray for Wide Vials For wide vials (28.5 x 95 mm) Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10149701
Estimated Shipment: 05-02-2027
Add to basket
Add to basket
Accommodate narrow- or wide-diameter vials.
  • Cardboard trays have cell dividers
  • 10 × 10 in. (25 x 25cm) row configuration
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) For wide vials (28.5 x 95 mm)
Includes Dividers
Material Cardboard
Type Tray for Wide Vials