missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AMG 900
GREENER_CHOICE

Product Code. p-200009250
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30000447

Brand: Tocris Bioscience™ 7865/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent pan-Aurora kinase inhibitor

AMG 900 is a potent pan-Aurora kinase inhibitor (IC50 values are 1 nM, 4 nM, and 5 nM for Aurora C, B, and A, respectively). It is more than 10-fold selective for Aurora kinases over JNK2, Tie2, p38α, Met, and TYK2. AMG 900 inhibits autophosphorylation of Aurora A and B, as well as the phosphorylation of histone H3 on Ser in HeLa cells. AMG 900 inhibits proliferation of 26 tumor cell lines (EC50 ranges from 0.7 nM - 5.3 nM), including Paclitaxel (Cat. No. 1097)-resistant tumor cell lines. In an HCT116 tumor xenograft model, AMG 900 inhibits phosphorylation of histone H3 and tumor growth. It is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-[[3-(2-Amino-4-pyrimidinyl)-2-pyridinyl]oxy]phenyl]-4-(4-methyl-2-thienyl)-1-phthalazinamine
CAS 945595-80-2
Quantity 10 mg
Target Aurora Kinase Inhibitors
Molecular Formula C28H21N7OS
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.