Learn More
Tocris Bioscience™ AM 4113
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AM 4113 is a high affinity and selective CB1 antagonist (Kd = 0.89 nM). Exhibits 100-fold selectivity for CB1 over CB2. Attenuates CB1 agonist AM 411-induced locomotor suppression and reduces food intake in vivo, but does not induce signs of nausea.
Specifications
Specifications
| Chemical Name or Material | 5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-1H-pyrazole-3-carboxamide |
| CAS | 614726-85-1 |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Molecular Formula | C17H12Cl3N3O |
| Formula Weight | Observed MW: 380.66 |