Learn More
Ansell AlphaTec™ 2000 STANDARD Jacket & Trouser Set Bound
Superior protection from low hazard liquid spray and fine particulates with bound seams for improved overall protection.
6975.00 NOK
Specifications
Material | Microporous Polyethylene Laminate (Non-woven) |
---|---|
Color | White |
Seam Type | Bound |
Certifications/Compliance (Europe) | Partial Body Protection Only, EN 13034:2005 + A1:2009, DIN 32781:2010, EN 14126:2003, ASTM F1671, EN ISO 13688:2013 |
Style | W20B219 |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17141015
|
Ansell
WH20B-00219-03 |
Medium |
6975.00 NOK
Pack of 40 |
Estimated Shipment: 23-12-2025
Log in to see stock. |
||||||
17151015
|
Ansell
WH20B-00219-04 |
Large |
6975.00 NOK
Pack of 40 |
Estimated Shipment: 23-12-2025
Log in to see stock. |
||||||
17161015
|
Ansell
WH20B-00219-05 |
X-Large |
6975.00 NOK
Pack of 40 |
Estimated Shipment: 23-12-2025
Log in to see stock. |
||||||
17171015
|
Ansell
WH20B-00219-06 |
2X-Large |
6975.00 NOK
Pack of 40 |
Estimated Shipment: 23-12-2025
Log in to see stock. |
||||||
Description
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Optimized fit, improving wearer comfort and safety
- Low Linting
Specifications
Microporous Polyethylene Laminate (Non-woven) | |
Bound | |
W20B219 |
White | |
Partial Body Protection Only, EN 13034:2005 + A1:2009, DIN 32781:2010, EN 14126:2003, ASTM F1671, EN ISO 13688:2013 | |
Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services |
Product Title
Ansell AlphaTec™ 2000 STANDARD Jacket & Trouser Set Bound