missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | AGAP2 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16863454
|
Proteintech
14690-1-AP-20UL |
20 ÎĽL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16853454
|
Proteintech
14690-1-AP-150UL |
150 ÎĽL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene belongs to the centaurin gamma-like family. It mediates anti-apoptotic effects of nerve growth factor by activating nuclear phosphoinositide 3-kinase. It is overexpressed in cancer cells, and promotes cancer cell invasion. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
AGAP2 | |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q3UHD9, Q8CGU4, Q99490 | |
116986, 216439, 65218 | |
AGAP2 Fusion Protein Ag6142 | |
Primary | |
-20°C | |
AGAP2 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AGAP 2, AGAP2, Centaurin gamma 1, CENTG1, Cnt g1, GGAP2, KIAA0167, PIKE | |
Agap2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AGAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech