Learn More
Thermo Scientific™ Acclaim™ C30 HPLC Columns
Shop All Thermo Scientific ProductsDescription
For separations of hydrophobic, structurally related isomers, Thermo Scientific™ Acclaim™ C30 LC reversed-phase columns provide superior results. The covalent modification of C30 alkyl silane on ultrapure, spherical, porous silica gel gives these columns high shape selectivity. They are fully compatible with various aqueous buffers, useful for a broad application range of analytes, and offer more flexibility in method development.
- Unique selectivity complementary to other reversed-phase columns
- High efficiency and rugged packing
- Extremely low bleed, fully compatible with MS
- Stable between pH 2 and 8
- Available in 3 and 5ÎĽm particle sizes and 4.6, 3.0, and 2.1mm diameters, with an average pore diameter of 200Ă…
Achieve the high standards set by modern HPLC and LC/MS methods with the Acclaim family of reversed-phase columns. These silica-based columns with advanced column bonding and endcapping technologies provide complementary selectivity, high column efficiencies, and ultra-high resolution for a wide variety of applications.
- Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases, and specialty phases including wide-pore, are available to meet your specific application needs
- Performance tested to ensure high quality and reliability for pharmaceutical, biopharmaceutical, environmental, food, and other industrial chromatographic separations
Specifications
Specifications
Column Format | Analytical Column |
Diameter (Metric) | 4.6 mm |
Length (Metric) | 250 mm |
Max. Pressure | 8000 psi (550 bar) |
Particle Size | 5 ÎĽm |
pH | 2 to 8 |
Pore Size | 200 Ă… |
Quantity | Each |
Surface Area | 200 m²/g |
Temperature | 60°C |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Documents
Scientific Resources
Application Notes
Posters
Product Information
Manuals
Specification Sheets