Learn More
Ohaus™ 6 x Centrifuge Tube Adapters, Open Bottom, Frontier™ Fixed Angle Rotors
Shop All Ohaus Products
Click to view available options
Capacity:
0.2/0.4 mL
0.5 mL
1.5/2.0 mL
Unit Size:
Pack of 6
Description
- Accommodates centrifuge tubes that are smaller than the space provided in the rotor carrier
- Flat-, open-bottom carrier.
- Suitable for use in the following Frontier™ rotors:
- R-A30x2/13MS
- R-A24x2/15MS
- R-A24x2/15M
- R-A12x2/18MI
- R-A24x2/16MI
- R-A30x2/14MIS
- R-A44x2/16MI
- R-A24x2/16MIS
Specifications
Specifications
Capacity | 0.2/0.4 mL |
Max. Tube Diameter | 6 mm |
For Use With (Equipment) | Frontier Fixed-Angle Rotors |
Description | Adapter, D6mm, 0.2/0.4ml Tubes x6 |
No. of Places | 1 |
No. per Pack | 6 |
Product Content Correction
Product Title
Ohaus™ 6 x Centrifuge Tube Adapters, Open Bottom, Frontier™ Fixed Angle Rotors >