Learn More
DiscoverX MDL 29,951
Click to view available options
:
5 mg
Description
MDL 29,951 is a synthetic small molecule selective agonist of the G protein-coupled receptor 17 (GPR17) that is active in intact cells. Selective over endogenous purinergic receptors. May be used in GPR17 studies and has a role in the maturation of oligodendrocytes. Provided as 5mg of dry powder.
Specifications
Specifications
| Quantity | 5 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.