Learn More
DiscoverX MDL 29,951
Product Code. 17352579
Encompass
Click to view available options
:
5 mg
Product Code. 17352579
Brand: DiscoverX 921264
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MDL 29,951 is an agonist for GPR17. Provided as 5mg of dry powder.
MDL 29,951 is a synthetic small molecule selective agonist of the G protein-coupled receptor 17 (GPR17) that is active in intact cells. Selective over endogenous purinergic receptors. May be used in GPR17 studies and has a role in the maturation of oligodendrocytes. Provided as 5mg of dry powder.
Specifications
Quantity 5 mg