missing translation for 'onlineSavingsMsg'
Learn More

Reagecon 5LT BUFFER SOLUTION PH11.00 0.05 20°C 5L

Brand:  Reagecon 5110

5LT BUFFER SOLUTION PH11.00 0.05 20°C 5L

Product Code. 11474529

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1122L1 Certificate TypeCertificate of Analysis Date20/12/2022 Catalog Number5110
Lot Number1122A1 Certificate TypeCertificate of Analysis Date24/11/2022 Catalog Number5110
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Reagecon 5LT BUFFER SOLUTION PH11.00 0.05 20°C 5L >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.