Learn More
Brannan™ ECO-therm Lab Thermometer
Product Code. 15412695
Click to view available options
Temperature Range (Metric):
-10°C to +110°C
-10°C to +150°C
-35°C to +50°C
Immersion Type:
Partial Immersion
Total Immersion
Unit Size:
Pack of 10
Product Code. 15412695
Brand: Brannan™ 46/706/1
Green spirit thermometers filled with non toxic expansion liquid.
  • Quality and resolution make these ideal substitute for general purpose mercury thermometers
  • Each thermometer is fitted with Brannan non roll end cap
  • Cap has integral ring to allow the thermometer to be suspended and is supplied in protective PVC packaging
Specifications
Immersion Type Total Immersion
Length (Metric) 305 mm
Material Glass
Thermometer Fill Green Spirit
Product Type Laboratory Immersion Thermometer
Max. Temperature (Metric) 150°C
Min. Temperature (Metric) -10°C
Temperature Range (Metric) -10°C to +150°C
Accuracy ±1.5°C
Subdivision 1°C