Learn More
3-(Methylthio)phenyl isothiocyanate, 97%, Thermo Scientific™
Product Code. 11307836
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307836 1 g 1g
11317836 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11307836 Supplier Thermo Scientific Alfa Aesar Supplier No. L12999.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51333-80-3
Molecular Formula C8H7NS2
Molecular Weight (g/mol) 181.271
MDL Number MFCD00041079
InChI Key IHKAGFVCXPFGRP-UHFFFAOYSA-N
Synonym 3-methylthio phenyl isothiocyanate, 1-isothiocyanato-3-methylsulfanyl benzene, 3-isothiocyanatophenyl methyl sulfane, 1-isothiocyanato-3-methylsulfanyl-benzene, acmc-20anif, 3-methylthiobenzenisothiocyanate, 3-methylthio phenylisothiocyanate, 3-methylthio-phenyl isothiocyanate, 3-methylthiophenyl isothiocyanate, 1-isothiocyanato-3-methylthio benzene
PubChem CID 142841
IUPAC Name 1-isothiocyanato-3-methylsulfanylbenzene
SMILES CSC1=CC=CC(=C1)N=C=S
Specifications
Density 1.22
Boiling Point 144°C to 145°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.691
Quantity 1 g
UN Number UN2922
Beilstein 2208144
Sensitivity Moisture sensitive
Formula Weight 181.28
Percent Purity 97%
Chemical Name or Material 3-(Methylthio)phenyl isothiocyanate