accessibility menu, dialog, popup

UserName

CH3SNCS

CAS RN 51333-80-3

IUPAC Name: 1-isothiocyanato-3-(methylsulfanyl)benzene
Synonyms: 1-isothiocyanato-3-(methylsulfanyl)benzene
Molecular Weight (g/mol): 181.27
Molecular Formula: C8H7NS2
InChi Key: IHKAGFVCXPFGRP-UHFFFAOYSA-N
SMILES: CSC1=CC=CC(=C1)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)