Specifications
| Quantity | 1 Each |
| Pore Size | 1.40 mm |
| Type | Sieve |
| Certifications/Compliance | Certificate of Conformity, BS 410: Part 1: 2000, ISO 3310: Part 1: 2000 |
| Material | Stainless Steel |
| Includes | Certificate of conformity and record card |
| Diameter (Metric) Outer | 200 mm |
| Frame Material | Stainless Steel |
| Mesh Material | Woven Wire |