Learn More
Thermo Scientific™ 2000mL BIOS Swinging Bucket Rotors
Shop All Thermo Scientific Products
Click to view available options
Capacity:
2000 mL
6 x 2000 mL
8 x 2000 mL
Includes:
Oval Bucket (set of 2)
Rotor, Oval Buckets, 2000 mL Bottles
Unit Size:
Each
Pack of 2
Description
Thermo Scientific™ 2000mL BIOS swinging bucket rotors are designed for use with the Thermo Scientific™ Sorvall™ BIOS 16 bioprocessing centrifuge. The rotor, buckets and bottles which are provided as a set, have been engineered to simplify operation of the centrifuge while maximizing productivity.
Achieve efficient and reliable sample processing with the following features:
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
- 6 x 2000mL (12L) capacity at 4,700 max rpm (7,187 x g max RCF) operation;
- 8 x 2000mL (16L) capacity at 3,900 max rpm (5,374 x g max RCF) operation
- Easy-to-use innovative 2000mL bucket and bottle with a wide mouth design requires no bridge and are resistant to 1000 cycles
- Thermo Scientific™ Eco-Spin™ windshield provides energy savings up to 60% when compared with non-windshielded rotors
- Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ enabling instant lid opening, storage and rotor identification that saves time and protects the integrity of your samples
- 2-year warranty
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Specifications
Specifications
Material | Stainless Steel Rotor Body, Aluminum Windshield and Bucket, Polypropylene Bottle |
Type | Swinging Bucket Rotor |
Capacity | 6 x 2000 mL |
Max. RCF | 7,187 x g |
Max. Speed | 4,700 rpm |
For Use With (Equipment) | Thermo Scientific Sorvall BIOS 16 Centrifuge |
Includes | Rotor, Oval Buckets, 2000 mL Bottles |
Documents
Product Information
Specification Sheets
8 x 2000mL BIOS Swinging Bucket Rotor RTBA Data Sheet [EN]
File size: 563.3 kB
6 x 2000mL BIOS Swinging Bucket Rotor RTBA Data Sheet [EN]
File size: 595.5 kB
Scientific Resources
Brochures
Sorvall BIOS 16 Centrifuge Brochure [EN]
File size: 1.9 MB
Product Content Correction
Product Title
Thermo Scientific™ 2000mL BIOS Swinging Bucket Rotors >