Learn More
Fisherbrand™ -20 to +110 Lo Tox™ Laboratory Immersion Thermometer
Product Code. 15422695
Change view
Click to view available options
Temperature Range (Metric):
-20°C to +110°C
Unit Size:
Pack of 10
Product Code. Temperature Range (Metric) unitSize
15422695 -20°C to +110°C Pack of 10
1 options
Product Code. 15422695
Brand: Fisherbrand™ 44/802/9
Lo Tox™ organic filling is specially chosen to achieve wide temperature span and provide the lowest practical levels of hazard.
  • Quality and legibility make these an ideal substitute for mercury thermometers
  • Fitted with Brannan non roll end cap which has ring to allow the thermometer to be suspended
Specifications
Immersion Type Partial Immersion
Length (Metric) 305 mm
Material Glass
Thermometer Fill Blue Organic Liquid
Product Type Laboratory Immersion Thermometer
Max. Temperature (Metric) 110°C
Min. Temperature (Metric) -20°C
Temperature Range (Metric) -20°C to +110°C
Accuracy ±1°C at 35°C
Subdivision 0.5°C