missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylthiophenol, 98%, Thermo Scientific™

Product Code. 11431187
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11431187

Brand: Thermo Scientific Alfa Aesar B24164.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4001-61-0
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00010020
InChI Key NHAUBUMQRJWWAT-UHFFFAOYSA-N
Synonym 2,5-dimethylthiophenol, 2,5-dimethyl thiophenol, benzenethiol, 2,5-dimethyl, 2,5-dimethylbenzene-1-thiol, 2,5-dimethylthiophenol;2,5-dimethylthiophenol;, p-xylenethiol, 2-mercapto-p-xylene, 2,5dimethylthiophenol, pubchem6811, 2,5-dimethyl-thiophenol
PubChem CID 77615
IUPAC Name 2,5-dimethylbenzenethiol
SMILES CC1=CC(=C(C=C1)C)S

Specifications

Density 1.02
Boiling Point 126°C to 127°C (50 mmHg)
Flash Point 83°C (181°F)
Odor Unpleasant
Refractive Index 1.5698
Quantity 25 g
UN Number UN2810
Beilstein 2040348
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 98%
Chemical Name or Material 2,5-Dimethylthiophenol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.