accessibility menu, dialog, popup

UserName

CAS RN 4001-61-0

CH3CH3S

CAS RN 4001-61-0

IUPAC Name: (2,5-dimethylphenyl)sulfanide
Synonyms: (2,5-dimethylphenyl)sulfanide
Molecular Weight (g/mol): 137.22
Molecular Formula: C8H9S
InChi Key: NHAUBUMQRJWWAT-UHFFFAOYSA-M
SMILES: CC1=CC=C(C)C([S-])=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)