missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ MRS 2905
Description
MRS 2905 is a potent and selective P2Y14 receptor agonist (EC50 = 0.92 nM). Exhibits >2000-fold selectivity over P2Y6.
Specifications
Specifications
| Chemical Name or Material | 2-Thiouridine-5′-O-(α, β-methylene)diphosphate trisodium salt |
| Quantity | 1 mg |
| Product Line | Tocris |
| Target | P2Y Receptor Agonists |
| Molecular Formula | C10H13N2Na3O10P2S |
| Formula Weight | 484.2 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?