Learn More
Tocris Bioscience™ PRX 07034
Description
PRX 07034 is a potent and selective 5-HT6 receptor antagonist (IC50 = 19 nM, Ki = 4 - 8 nM). Enhances working memory in rats.
Specifications
Specifications
| Chemical Name or Material | N-(1-(5-Chloro-2,3-dimethoxyphenyl)ethyl)-2-(methylsulfonyl)-5-(piperazin-1-yl)aniline hydrochloride |
| CAS | 903580-39-2 |
| Quantity | 10 mg |
| Molecular Formula | C21H28ClN3O4S.HCl |
| Formula Weight | 490.44 |
| Purity | 98% |