missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MLS 1547

Description
MLS 1547 is a G protein-biased D2 receptor partial agonist (Ki = 1.2 μM). Stimulates D2 receptor G protein-mediated signaling and acts as an antagonist of dopamine-mediated β-arrestin recruitment at D2 receptors.
Specifications
Specifications
| Chemical Name or Material | 5-Chloro-7-[[4-(2-pyridinyl)-1-piperazinyl]methyl]-8-quinolinol |
| CAS | 315698-36-3 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C19H19ClN4O |
| Formula Weight | Observed MW: 354.83 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?