Learn More
Tocris Bioscience™ DFHO
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
DFHO is a mimic of red fluorescent protein (RFP) fluorophore for imaging RNA in living cells. DFHO fluorescence is activated by binding to Corn aptamer (Kd = 70 nM). Excitation/emission maxima (λ) = 505/545 nm; extinction coefficient = 29000 M-1 cm-1.
Specifications
Specifications
| Chemical Name or Material | 4-(3,5-Difluoro-4-hydroxybenzylidene)-1-methyl-5-oxo-4,5-dihydro-1H-imidazole-2-carbaldehyde oxime |
| CAS | 1420815-34-4 |
| Quantity | 10 mg |
| Target | Light-up Aptamers |
| Molecular Formula | C12H9F2N3O3 |
| Formula Weight | 281.22 |
| Product Type | Fluorophore |
| Purity | 98% |