CAS RN 987-65-5
CAS RN 987-65-5
IUPAC Name:
disodium hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate
Synonyms:
disodium adenosine triphosphate
Molecular Weight (g/mol):
551.15
Molecular Formula:
C10H14N5Na2O13P3
InChi Key:
TTWYZDPBDWHJOR-IDIVVRGQSA-L
SMILES:
[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
1
–
2
of
2
results
2
ATP disodium salt, MedChemExpress
MedChemExpress ATP disodium salt (Adenosine 5'-triphosphate disodium salt) is a central component of energy storage and metabolism in vivo, provides the metabolic energy to drive metabolic pumps and serves as a coenzyme in cells. ATP disodium salt is an important endogenous signaling molecule in immunity and inflammation.
