CAS RN 978-62-1
CAS RN 978-62-1
IUPAC Name:
N-[3,5-bis(trifluoromethyl)phenyl]-5-chloro-2-hydroxybenzamide
Synonyms:
N-[3,5-bis(trifluoromethyl)phenyl]-5-chloro-2-hydroxybenzamide
Molecular Weight (g/mol):
383.67
Molecular Formula:
C15H8ClF6NO2
InChi Key:
CHILCFMQWMQVAL-UHFFFAOYSA-N
SMILES:
OC1=C(C=C(Cl)C=C1)C(=O)NC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
1
–
3
of
3
results
2
Tocris Bioscience™ IMD 0354
CAS: 978-62-1 Molecular Formula: C15H8ClF6NO2
3
IMD-0354, MedChemExpress
MedChemExpress IMD-0354 (IKK2 Inhibitor V) is a selective IKKβ inhibitor which inhibits NF-κB activity. IMD0354 inhibits TNF-α induced NF-κB transcription activity with an IC50 of 1.2 uM.