accessibility menu, dialog, popup

Test

CAS RN 942183-80-4

CAS RN 942183-80-4

IUPAC Name: 1-(2-oxo-2-{4-[4-(pyrimidin-2-yl)phenyl]piperazin-1-yl}ethyl)-N-[3-(pyridin-4-yl)-1H-indazol-5-yl]pyrrolidine-3-carboxamide
Synonyms: 1-(2-oxo-2-{4-[4-(pyrimidin-2-yl)phenyl]piperazin-1-yl}ethyl)-N-[3-(pyridin-4-yl)-1H-indazol-5-yl]pyrrolidine-3-carboxamide
Molecular Weight (g/mol): 587.69
Molecular Formula: C33H33N9O2
InChi Key: HDAJDNHIBCDLQF-UHFFFAOYNA-N
SMILES: O=C(CN1CCC(C1)C(=O)NC1=CC=C2NN=C(C2=C1)C1=CC=NC=C1)N1CCN(CC1)C1=CC=C(C=C1)C1=NC=CC=N1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)