accessibility menu, dialog, popup

UserName

CAS RN 93602-66-5

CH3NHH3CH3CONNNNNH2HOOPSOO

CAS RN 93602-66-5

IUPAC Name: triethylazanium 6-(6-amino-9H-purin-9-yl)-7-hydroxy-2-sulfanylidene-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Synonyms: TEAH 6-(6-aminopurin-9-yl)-7-hydroxy-2-sulfanylidene-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate
Molecular Weight (g/mol): 446.46
Molecular Formula: C16H27N6O5PS
InChi Key: OXIPZMKSNMRTIV-UHFFFAOYNA-N
SMILES: CC[NH+](CC)CC.NC1=C2N=CN(C3OC4COP([O-])(=S)OC4C3O)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results