accessibility menu, dialog, popup

UserName

CAS RN 926023-82-7

CH3OONH2NSHCl

CAS RN 926023-82-7

IUPAC Name: N-(3-aminopropyl)-2-[(3-methylphenyl)methoxy]-N-[(thiophen-2-yl)methyl]benzamide hydrochloride
Synonyms: N-(3-aminopropyl)-2-[(3-methylphenyl)methoxy]-N-(thiophen-2-ylmethyl)benzamide hydrochloride
Molecular Weight (g/mol): 430.99
Molecular Formula: C23H27ClN2O2S
InChi Key: UDXGBANGPYONOK-UHFFFAOYSA-N
SMILES: Cl.CC1=CC(COC2=CC=CC=C2C(=O)N(CCCN)CC2=CC=CS2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results