accessibility menu, dialog, popup

UserName

CAS RN 91917-65-6

CH3OONNNCH3ONNN

CAS RN 91917-65-6

IUPAC Name: ethyl 12-azido-8-methyl-9-oxo-2,4,8-triazatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaene-5-carboxylate
Synonyms: ethyl 12-azido-8-methyl-9-oxo-2,4,8-triazatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaene-5-carboxylate
Molecular Weight (g/mol): 326.32
Molecular Formula: C15H14N6O3
InChi Key: CFSOJZTUTOQNIA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C2CN(C)C(=O)C3=CC(=CC=C3N2C=N1)N=[N+]=[N-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results