CAS RN 912545-86-9
IUPAC Name:
3-chloro-N-[4-(methylamino)cyclohexyl]-N-{[3-(pyridin-4-yl)phenyl]methyl}-1-benzothiophene-2-carboxamide
Synonyms:
smoothened agonist
Molecular Weight (g/mol):
490.06
Molecular Formula:
C28H28ClN3OS
InChi Key:
VFSUUTYAEQOIMW-UHFFFAOYSA-N
SMILES:
CNC1CCC(CC1)N(CC1=CC(=CC=C1)C1=CC=NC=C1)C(=O)C1=C(Cl)C2=CC=CC=C2S1
1
–
3
of
3
results
2
SAG, MedChemExpress
MedChemExpress SAG is a potent Smoothened (Smo) receptor agonist (EC50=3 nM; Kd=59 nM). SAG activates the Hedgehog signaling pathway and counteracts Cyclopamine (HY-17024) inhibition of Smo.
3