CAS RN 90259-31-7
CAS RN 90259-31-7
IUPAC Name:
2-bromo-6-methylbenzoic acid
Synonyms:
2-bromo-6-methylbenzoic acid
Molecular Weight (g/mol):
215.05
Molecular Formula:
C8H7BrO2
InChi Key:
ICXBPDJQFPIBSS-UHFFFAOYSA-N
SMILES:
CC1=C(C(O)=O)C(Br)=CC=C1
1
–
1
of
1
results
1
2-Bromo-6-methylbenzoic acid, 98%
CAS: 90259-31-7 Molecular Formula: C8H7BrO2 Molecular Weight (g/mol): 215.05 MDL Number: MFCD01310788 InChI Key: ICXBPDJQFPIBSS-UHFFFAOYSA-N Synonym: 6-bromo-o-toluic acid,2-bromo-6-methyl-benzoic acid,6-bromo-2-methylbenzoic acid,benzoic acid, 2-bromo-6-methyl,bromtoluylsaure,bromotoluic acid,pubchem4715,acmc-209r5e,ksc494k9r,3-bromo-2-carboxytoluene PubChem CID: 2735589 IUPAC Name: 2-bromo-6-methylbenzoic acid SMILES: CC1=C(C(O)=O)C(Br)=CC=C1