Learn More
2-Bromo-6-methylbenzoic acid, 98%
Product Code. 11364596
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 11364596
Brand: Thermo Scientific Alfa Aesar L20149.06
Request Bulk or Custom Format
CAS: 90259-31-7 | C8H7BrO2 | 215.05 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 90259-31-7
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD01310788
InChI Key ICXBPDJQFPIBSS-UHFFFAOYSA-N
Synonym 6-bromo-o-toluic acid, 2-bromo-6-methyl-benzoic acid, 6-bromo-2-methylbenzoic acid, benzoic acid, 2-bromo-6-methyl, bromtoluylsaure, bromotoluic acid, pubchem4715, acmc-209r5e, ksc494k9r, 3-bromo-2-carboxytoluene
PubChem CID 2735589
IUPAC Name 2-bromo-6-methylbenzoic acid
SMILES CC1=C(C(O)=O)C(Br)=CC=C1
Specifications
Melting Point 107°C to 111°C
Quantity 5 g
Beilstein 2614094
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-methylbenzoic acid