accessibility menu, dialog, popup

UserName

CAS RN 885101-89-3

OHNOOH

CAS RN 885101-89-3

IUPAC Name: 3-(4-{[(3-phenoxyphenyl)methyl]amino}phenyl)propanoic acid
Synonyms: 3-(4-{[(3-phenoxyphenyl)methyl]amino}phenyl)propanoic acid
Molecular Weight (g/mol): 347.41
Molecular Formula: C22H21NO3
InChi Key: DGENZVKCTGIDRZ-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C(NCC2=CC(OC3=CC=CC=C3)=CC=C2)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results