CAS RN 883065-90-5
CAS RN 883065-90-5
IUPAC Name:
4-(2,3-dihydro-1,4-benzodioxin-6-yl)-3-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-4,5-dihydro-1H-1,2,4-triazol-5-one
Synonyms:
4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-2H-1,2,4-triazol-3-one
Molecular Weight (g/mol):
379.37
Molecular Formula:
C13H9N5O5S2
InChi Key:
QFRLDZGQEZCCJZ-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CN=C(SC2=NNC(=O)N2C2=CC3=C(OCCO3)C=C2)S1
1
–
2
of
2
results
1
Tocris Bioscience™ BI 78D3
CAS: 883065-90-5 Molecular Formula: C13H9N5O5S2
2
BI-78D3, MedChemExpress
MedChemExpress BI-78D3 functions as a substrate competitive inhibitor of JNK, inhibit the JNK kinase activity (IC50=280 nM).