accessibility menu, dialog, popup

UserName

CAS RN 75626-17-4

IUPAC Name: 1,4-difluoro-2-methoxybenzene
Synonyms: 1,4-difluoro-2-methoxybenzene
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: HUDMAQLYMUKZOZ-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC=C1F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results