missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoroanisole, 98%, Thermo Scientific™

Product Code. 11482387
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482387 100 g 100g
11462387 5 g 5g
11472387 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482387 Supplier Thermo Scientific Alfa Aesar Supplier No. A14255.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75626-17-4
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00042561
InChI Key HUDMAQLYMUKZOZ-UHFFFAOYSA-N
PubChem CID 2724989
IUPAC Name 1,4-difluoro-2-methoxybenzene
SMILES COC1=C(C=CC(=C1)F)F

Specifications

Boiling Point 72°C to 73°C (25 mmHg)
Flash Point 40°C (104°F)
Refractive Index 1.4755
Quantity 100 g
UN Number UN1993
Beilstein 4383722
Synonym 2,5-difluoroanisole, 2,5-difluoromethoxybenzene, benzene, 1,4-difluoro-2-methoxy, 2,5-difluoro-1-methoxybenzene, 2,5-difluoroanisol, pubchem3456, acmc-209oz7, ksc641o8p, 1,4-difluoro-2-methoxy-benzene, benzene,1,4-difluoro-2-methoxy
Formula Weight 144.12
Percent Purity 98%
Chemical Name or Material 2,5-Difluoroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.