accessibility menu, dialog, popup

UserName

CAS RN 63597-44-4

H3COO(R)(R)CH3(S)OH(S)OH(S)OH(S)CH3O(R)H3CCH3(S)OH3CO

CAS RN 63597-44-4

IUPAC Name: (1S,2S,6S,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Synonyms: (1S,2S,6S,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Molecular Weight (g/mol): 616.84
Molecular Formula: C36H56O8
InChi Key: PHEDXBVPIONUQT-LQLWEASQSA-N
SMILES: CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results