CAS RN 635728-49-3
CAS RN 635728-49-3
IUPAC Name:
(3R,3aS,6aR)-hexahydrofuro[2,3-b]furan-3-yl N-[(2S,3R)-3-hydroxy-4-[N-(2-methylpropyl)4-aminobenzenesulfonamido]-1-phenylbutan-2-yl]carbamate; ethanol
Synonyms:
darunavir; ethyl alcohol
Molecular Weight (g/mol):
593.74
Molecular Formula:
C29H43N3O8S
InChi Key:
QWSHKNICRJHQCY-VBTXLZOXSA-N
SMILES:
CCO.CC(C)CN(C[C@@H](O)[C@H](CC1=CC=CC=C1)NC(=O)O[C@H]1CO[C@H]2OCC[C@@H]12)S(=O)(=O)C1=CC=C(N)C=C1
1
–
3
of
3
results
2
Darunavir Ethanolate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Darunavir Ethanolate, MedChemExpress
MedChemExpress Darunavir ethanolate (TMC114 Ethanolate) is a potent HIV protease inhibitor used to treat and prevent HIV/AIDS. Darunavir has a Ki of 1 nM for wild type HIV-1 protease.