CAS RN 63466-71-7
CAS RN 63466-71-7
IUPAC Name:
pentacyclo[10.6.2.0²,⁷.0⁹,¹⁹.0¹⁶,²⁰]icosa-1(18),2,4,6,8,10,12,14,16,19-decaene
Synonyms:
benzopyrene benzo(d,e,f)chrysene 3,4-benzopirene 3,4-benzo(a)pyrene 3,4 benzopyrene 3,4-benzpyren 3,4 benzpyrene BaP benzo(A)pyrene 6,7-benzopyrene
Molecular Weight (g/mol):
252.32
Molecular Formula:
C20H12
InChi Key:
FMMWHPNWAFZXNH-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=CC=C2C1=C34
1
–
4
of
4
results
3
Deuterated PAH Mixture 189 10 μg/mL in Toluene, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
4
Benzo[a]pyrene-d12, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.