Learn More
Benzo[a]pyrene-d12, CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Benzo(a)pyrene D12 |
| CAS | 63466-71-7 |
| Molecular Formula | C20 2H12 |
| Quantity | 0.01 g |
| Synonym | Benzo[a]pyrene-1,2,3,4,5,6,7,8,9,10,11,12-d12, Benzo[a]pyrene-d12 (9 CI), Benzo[a]pyrene d12, Perdeuterated benzo[a]pyrene, [12-2 H]Benzo[a]pyrene, Benzo[a]pyrene D12 |
| InChI Formula | InChI=1 S/C20H12/c1-2-7-17-15(4-1)12-16-9-8-13-5-3-6-14-10-11-18(17)20(16)19(13)14/h1-12 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c([2 H])c3c([2 H])c([2 H])c4c([2 H])c([2 H])c([2 H])c5c([2 H])c([2 H])c2c3c45 |
| IUPAC Name | 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuteriobenzo[a]pyrene |
| Molecular Weight (g/mol) | 264.38 |
| Formula Weight | 264.1692 g/mol |
| Show More |