Learn More
Deuterated PAH Mixture 189 10 μg/mL in Toluene, Dr. Ehrenstorfer
Click to view available options
Quantity:
5x1 mL
Unit Size:
5mL
Specifications
Specifications
| Chemical Name or Material | Benzo(a)pyrene D12∣Benzo(b)fluoranthene D12∣Benzo[a]anthracene D12∣Chrysene D12 |
| CAS | 63466-71-7, 93951-98-5, 1718-53-2, 1719-03-5 |
| Solvent or Matrix | Toluene |
| Quantity | 5x1 mL |
| Concentration or Composition (by Analyte or Components) | Benzo(b)fluoranthene D12 9.868 μg/mL∣Benzo(a)pyrene D12 10.02 μg/mL∣Chrysene D12 10.04 μg/mL∣Benzo[a]anthracene D12 10.09 μg/mL |