accessibility menu, dialog, popup

UserName

CAS RN 5460-79-7

CH3ONH3CN

CAS RN 5460-79-7

IUPAC Name: 4-(3-methoxyphenyl)-1-methylpiperidine-4-carbonitrile
Synonyms: 4-(3-methoxyphenyl)-1-methylpiperidine-4-carbonitrile
Molecular Weight (g/mol): 230.31
Molecular Formula: C14H18N2O
InChi Key: COYLNHITVXBZPK-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C1(CCN(C)CC1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)