Learn More
4-(3-Methoxyphenyl)-1-methyl-4-piperidinecarbonitrile, TRC
Product Code. 30474417
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474417 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30474417 Supplier Toronto Research Chemicals Supplier No. TRCM2661951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(3-Methoxyphenyl)-1-methyl-4-piperidinecarbonitrile
CAS 5460-79-7
Molecular Formula C14H18N2O
Quantity 1 g
Synonym 4-(3-Methoxyphenyl)-1-methylpiperidine-4-carbonitrile, 4-(m-Methoxyphenyl)-1-methylisonipecotonitrile, 4-Cyano-4-m-methoxyphenyl-1-methylpiperidine, NSC 21330
InChI Formula InChI=1S/C14H18N2O/c1-16-8-6-14(11-15,7-9-16)12-4-3-5-13(10-12)17-2/h3-5,10H,6-9H2,1-2H3
SMILES COc1cccc(c1)C2(CCN(C)CC2)C#N
IUPAC Name 4-(3-methoxyphenyl)-1-methylpiperidine-4-carbonitrile
Molecular Weight (g/mol) 230.31
Formula Weight 230.1419