Learn More
4-(3-Methoxyphenyl)-1-methyl-4-piperidinecarbonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(3-Methoxyphenyl)-1-methyl-4-piperidinecarbonitrile |
| CAS | 5460-79-7 |
| Molecular Formula | C14H18N2O |
| Quantity | 100 mg |
| Synonym | 4-(3-Methoxyphenyl)-1-methylpiperidine-4-carbonitrile, 4-(m-Methoxyphenyl)-1-methylisonipecotonitrile, 4-Cyano-4-m-methoxyphenyl-1-methylpiperidine, NSC 21330 |
| InChI Formula | InChI=1S/C14H18N2O/c1-16-8-6-14(11-15,7-9-16)12-4-3-5-13(10-12)17-2/h3-5,10H,6-9H2,1-2H3 |
| SMILES | COc1cccc(c1)C2(CCN(C)CC2)C#N |
| IUPAC Name | 4-(3-methoxyphenyl)-1-methylpiperidine-4-carbonitrile |
| Molecular Weight (g/mol) | 230.31 |
| Formula Weight | 230.1419 |