accessibility menu, dialog, popup

UserName

CAS RN 52205-73-9

NaNaCH3(S)(S)(R)OONClCl(S)(S)OPOOO

CAS RN 52205-73-9

IUPAC Name: disodium (1S,3aS,3bR,9bS,11aS)-7-{[bis(2-chloroethyl)carbamoyl]oxy}-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl phosphate
Synonyms: emcyt estracyt estramustine phosphate sodium estramustine sodium phosphate estramustine phosphate disodium
Molecular Weight (g/mol): 564.35
Molecular Formula: C23H30Cl2NNa2O6P
InChi Key: IIUMCNJTGSMNRO-VVSKJQCTSA-L
SMILES: [Na+].[Na+].C[C@]12CC[C@H]3[C@@H](CCC4=CC(OC(=O)N(CCCl)CCCl)=CC=C34)[C@@H]1CC[C@@H]2OP([O-])([O-])=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results